missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Water, for qPCR or RT-qPCR, Fisher BioReagents
High purity water rigorously tested for degrading enzymes and genomic DNA that interefere with amplification and can lead to false-positive signals in PCR, qPCR and RT-qPCR applications.
$424.63 - $1477.68
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP28251
|
Thermo Fisher Scientific
BP28251 |
1 L | Poly Bottle |
Each for $424.63
Case of 6 Each for $2,547.78
|
|
||||
BP282510
|
Thermo Fisher Scientific
BP282510 |
10 L | Poly CUBE |
Each for $1,477.68
|
|
||||
Description
- Minimize the risk of false positive signals
- Tested for DNase, RNase, protease, nickase, and E. coli and human genomic DNA using sensitive assays
- Evaluated for metal impurities at low ppb concentrations to ensure optimal enzymatic activity, especially for polymerase during PCR
- Sub-micron filtered to remove particulates
- Suitable for use in qPCR and RT-qPCR for genomics work, microarrays, and other sensitive life science applications
- Also useful for the manufacturing of PCR-related kits and mastermixes
Chemical Identifiers
7732-18-5 | |
18.015 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Specifications
5.4 to 7.0 | |
7732-18-5 | |
100°C | |
Not detected | |
<0.1EU/mL | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Poly Bottle | |
Liquid |
Clear, Colorless | |
0°C | |
≤2μS/cm | |
H2O | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
qPCR | |
1 L |
Safety and Handling
Recommended Storage : 4°C; Ships at ambient temperature