missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Clear Polystyrene 96-Well Microplates
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
$770.00 - $1321.88
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Binding Type | Not Treated |
Color | Clear |
Material | Polystyrene |
Catalog Number | Mfr. No. | Surface Treatment | Sterility | Well Shape | Lid | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Surface Treatment | Sterility | Well Shape | Lid | Price | Quantity | ||
07-200-760
|
Corning™
3788 |
Non-treated | Sterile | Round | With Lid |
Case of 100 Each for $1,105.51
|
|
||
07-200-107
|
Corning™
C3896 |
Non-treated | Sterile | Conical Bottom (V-well) | Without Lid |
Case of 48 Each for $939.41
|
|
||
07-200-106
|
Corning™
3798 |
Non-treated | Non-sterile | Round | Without Lid |
Case of 100 Each for $1,129.37
|
|
||
07-200-109
|
Corning™
3898 |
Treated | Non-sterile | Conical Bottom (V-well) | Without Lid |
Case of 100 Each for $1,321.88
|
|
||
07-200-704
|
Corning™
3367 |
Non-treated | Sterile | Round | Without Lid |
Case of 50 Each for $770.00
|
|
||
07-200-103
|
Corning™
3795 |
Non-treated | Sterile | Round | Without Lid |
Case of 100 Each for $898.99
|
|
||
07-200-108
|
Corning™
C3897 |
Non-treated | Non-sterile | Conical Bottom (V-well) | Without Lid |
Case of 100 Each for $902.40
|
|
||
07-200-656
|
Corning™
3370 |
Non-treated | Sterile | Flat Bottom (F-well) | With Lid |
Case of 100 Each for $1,165.71
|
|
||
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Standard | |
Not Treated | |
Polystyrene | |
75 to 200 μL |
8 x 12 | |
Clear | |
96 |