missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Denatured Alcohol, 95%, Fisherbrand™, HistoPrep™
Supplier: Epredia HC11001GL
Specifications
Liquid | |
-114.1°C | |
64-17-5,67-56-1,7732-18-5 | |
Poly Bottle | |
Clear | |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
propan-2-ol | |
702 | |
Isopropyl alcohol, Ethyl alcohol, Methyl alcohol, Water |
Denatured Alcohol, 95% | |
78.5°C | |
0.822g/cm³ | |
40mmHg | |
1 gal. | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
46.07 | |
CHEBI:16236 |
Chemical Identifiers
64-17-5 | |
46.07 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:16236 | |
CC(C)O |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
propan-2-ol |