DuPont™ MOLYKOTE™ High-Vacuum Grease
Designed for sealing and lubricating vacuum and pressure systems.
Supplier:  DuPont™ 1658832
 View more versions of this product
Catalog No. 14-635-5C
Explore available promotions
Description
Description
Designed for sealing and lubricating vacuum and pressure systems
  • Low vapor pressure, stiff, nonmelting silicone lubricant maintains its consistency from -40° to +204°C (-40° to +400°F)
  • Resists oxidation and chemicals; noncarbonaceous
Specifications
Specifications
High Vacuum Grease
FDA 21 CFR 175.300, USDA, NSF 61
Silicone
8 lb.
-40°C to +204°C
Designed for sealing and lubricating vacuum and pressure system
White
Jar
-40°F to 400°F
2 lb.
Product Suggestions
Product Suggestions