missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals FastDNA™ SPIN Kit for Soil
FastDNA™ SPIN Kit for Soil is used for rapid isolation of PCR-ready genomic DNA from any environmental sample.
$649.49 - $1591.33
Specifications
Capacity | 2 mL |
---|---|
Concentration | Up to 500 mg per prep |
Format | Kit |
Isolation Technology | Glass Milk |
Sample Type | Soil, Feces, Sludge, Wastewater, Environmental, Compost, Manure, Rhizosphere |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
MP116560200
|
MP Biomedicals
116560200 |
50 Preps |
Each for $649.49
|
|
|||||
MP116560300
|
MP Biomedicals
116560300 |
100 Preps |
Each for $1,591.33
|
|
|||||
Description
- The FastDNA™ SPIN Kit for Soil quickly and efficiently isolates bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples.
- Soil is processed by a FastPrep™ instrument and Lysing Matrix E tubes designed to efficiently lyse all microorganisms.
- The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications.
- Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
- The FastDNA Spin Kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediments, sludge, compost, manure, rhizosphere or waste water.
- The FastDNA Spin Kit for Soil contains Lysing Matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors.
Specifications
2 mL | |
Kit | |
Soil, Feces, Sludge, Wastewater, Environmental, Compost, Manure, Rhizosphere | |
Isolate bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples, including sediment, sludge, compost, manure, rhizosphere, waste water and more | |
Isolate bacterial, fungal, plant, and animal genomic DNA | |
30 min. |
Up to 500 mg per prep | |
Glass Milk | |
FastPrep™ Instrument | |
Nucleic Acid Isolation, Mini-prep | |
DNA |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.