Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors
Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.
$65.70 - $113.11
Catalog Number | Mfr. No. | Product Type | Length (Metric) | Temperature Range (English) | Temperature Range (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Length (Metric) | Temperature Range (English) | Temperature Range (Metric) | Price | Quantity | ||
15-077-20
|
N/A | Digital Thermometer: Dual Scale | 102 mm | -58°F to +158°F | -50°C to +70°C |
Each for $65.70
|
|
||
15-077-27
|
N/A | Digital Thermometer: Celsius Scale | 102 mm | N/A | -50°C to +70°C |
Each for $76.27
|
|
||
15-077-8D
|
N/A | Digital Thermometer: Dual Scale | 70 mm | -58°F to +158°F | -50°C to +70°C |
Each for $113.11
|
|
||
Description
Choose from models with a variety of features including MAX/MIN function to record the maximum and minimum temperature in a certain time period; visual and audible alarms and multiple displays of recorded temperatures.
- Select the correct temperature to use in water baths, refrigerators, incubators and tanks
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™ indicates traceability to standards provided by NIST™.
Specifications
±1°C | |
Digital LCD |