missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Bulb Pipette Filler, Red
Fill any standard size pipet without the risk of cracking or chipping. Fisherbrand™ Safety Bulb Pipet Filler uses an ABS attachment with a silicone adapter that seals around pipets without twisting. Comes with an elastic band for convenient hanging on bottles or wrist.
Includes: Handy elastic band for hanging on wrist or reagent bottle
Description
Uses an ABS attachment with a silicone adapter that seals around pipets without twisting. Includes an elastic band for convenient hanging on bottles or wrist. Allows you to fill any standard size pipet without the risk of cracking or chipping.
- Pipette does not contact hard plastic, which reduces the risk of cracking and chipping
- 15mL capacity, 106mmL x 41mmOD (4.5-6.5mm hole dia.)
- Elastic band for hanging on reagent bottle or wrist
- Filler fits standard size pipettes
Specifications
4.5-6.5mm | |
Pipette | |
Bulb | |
Pipette Filler | |
Pipet | |
3 Pack | |
107 mm |
Red with White Attachment | |
Rubber with ABS attachment | |
1 | |
15 mL | |
Handy elastic band for hanging | |
4.21 in. |