missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methanol, Optima™ for HPLC, Fisher Chemical™

High purity mobile phase for HPLC

Supplier:  Fisher Chemical A454SK4

 View more versions of this product

Catalog No. A454SK-4



Explore available promotions

Description

Description

High purity mobile phase for HPLC

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Mobile phase for analytical and preparative separations
  • Also meets ACS specifications
  • Intended for research use only
Specifications

Specifications

Methanol
67-56-1
Colorless
64.7°C
Pass Test
Liquid
1ppm max.
0.0003mEq/g max.
0.0002mEq/g max.
MFCD00004595
0.55 cP at 20°C
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
≥99.9%
-98°C
0.01 AU max. at 254nm, 0.11 AU max. at 230nm, 0.23 AU max. at 220nm, 1.00 AU max. at 205nm
0.791g/cm³
4 L
Safe-Cote™ Amber Glass
Also meets ACS specifications
99.9% min by GC
CH4O
CH3OH
128 hPa at 20°C
0.07% max.
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
OPTIMA™

Chemical Identifiers

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions