Minigrip™ SPECI-ZIP™ Reclosable Clear Biohazard Bags with Black and Orange Biohazard Warning
Designed with temperature indicators and tear line
 View more versions of this product
Catalog No. 22-310-405
Explore available promotions
Description
Description
Designed with temperature indicators and tear line
  • Double pocket, three-wall bags
  • Bilingual in English and Spanish
Specifications
Specifications
Reclosable Biohazard Bag
Easy-to-open tear line
12 in.
15 in.
Clear
Black and orange biohazard warning, double pocket, temperature indicators and tear line
30.48 cm
28.1 cm