Nasco Swing Samplers
Specially designed for collecting samples from a horizontal flowing stream.
Supplier:  Nasco B01366
 View more versions of this product
Catalog No. 01-812-91
Explore available promotions
Description
Description
Specially designed for collecting samples from a horizontal flowing stream
  • The end of the sampler swings, allowing collection from different angles, including 90°
  • The 6-ft. (1.83 m) adjustable fiberglass pole extends to 12 ft. (3.66 m) providing a long length for any sampling situation.
  • Unit comes with a 960 mL polyethylene bottle and the cover has a vinyl liner for leak-proof protection. The bottle is attached to a polyurethane holder and is held in place with a plastic snapper ring that has an adjustable locking device.
Specifications
Specifications
Swing Arm Sampler
960 mL polyethylene bottle (attaches to pole with plastic snapper ring), cover with vinyl liner
8 ft. pole, extendable to 24 ft.
Fiberglass
Allowing collection of samples from different angles