Cytiva Protein G Sepharose™ 4 Fast Flow Media
Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.
Supplier:  Cytiva 17061802
 View more versions of this product
Catalog No. 45-000-117
Explore available promotions
Description
Description
  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • High capacity, >20mg human IgG/mL medium; high flowrate
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of diagnostic and research applications
Specifications
Specifications
2mg Protein G/mL Medium
90 μm
25 mL
Used in a range of diagnostic and research applications
4% Cross Linked Agarose
3 to 9
2°C to 8°C
Recombinant Protein G lacking albumin binding region