missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ PYREX™ Screw Cap Culture Tubes with PTFE Lined Phenolic Caps

PTFE interface is firmly bonded to glued-in rubber liner

Supplier:  Corning™ 982616X

 View more versions of this product

Catalog No. 14-933-1A



Explore available promotions

Description

Description

Similar to Cat. No. 9825, but phenolic cap has a PTFE interface firmly bonded to the glued-in rubber liner, providing a sure and highly inert sealing face.

For phenolic caps, see Cat. No. 9998.

 

Specifications

Specifications

Black
Threaded
125 mm
Marking Spot
15-415
No
Screw Cap
Phenolic
15 mL
Borosilicate Glass
PTFE
Non-sterile
16 mm
Culture Tube
SDS
Documents

Documents

Promotions

Promotions