missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Research Grade Fetal Bovine Serum, Canadian Sourced

Fisherbrand™ Research Grade Fetal Bovine Serum is sourced from Canada and triple 0.1 μm filtered.

$616.85 - $616.85

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG – Nephelometer
Content And Storage ≤ -10°C
Country of Origin Canada
Form Liquid
Grade Research
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FB12999102
View Documents Promotion Details
Fisherbrand™
SH3A764803
500 mL
Each for $616.85
 
Description

Description

  • Canadian origin
  • Compliant with current good manufacturing practices (cGMP)
  • For research use only
Specifications

Specifications

Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGPT), Glutamic Oxaloacetic Transaminase (SGOT), pH, Total Protein (Ph. Eur. 2.5.33), Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality (Ph. Eur. 2.2.35), Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, Glucose, IgG – Nephelometer
Canada
Research
No
No
≤ -10°C
Liquid
Yes
< 35 mg/dL
SDS
missing translation for 'documents'

missing translation for 'documents'