Learn More
Thermo Scientific™ SuperSignal™ West Femto Maximum Sensitivity Substrate
Ultra-sensitive enhanced chemiluminescent (ECL) HRP substrate for low-femtogram-level detection by Western blot analysis.
$899.82 - $3817.22
Specifications
Product Type | Substrate |
---|---|
Detection Method | Chemiluminescent |
Form | Liquid |
Product Line | SuperSignal™ |
Substrate | Horseradish Peroxidase |
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity | ||||
FER34095X4
|
Thermo Scientific™
34095X4 |
Luminol/Enhancer, 50 mL | 4 x 100 mL |
Each for $2,318.70
|
|
||||
PI34095
|
Thermo Scientific™
LSG34095 |
Luminol/Enhancer, 50mL, Stable Peroxide Buffer, 50mL | 100 mL |
Each for $899.82
|
|
||||
FER34096X4
|
Thermo Scientific™
34096X4 |
Stable Peroxide Buffer, 100 mL | 4 x 200 mL |
Each for $3,817.22
|
|
||||
PI-34096
|
Thermo Scientific™
34096 |
Stable Peroxide Buffer, 100mL, Luminol/Enhancer, 100mL | 200 mL |
Each for $1,445.13
|
|
||||
Description
Thermo Scientific™ SuperSignal West Femto Substrate is an ultra-sensitive enhanced chemiluminescent substrate (ECL substrate) for detection of peroxidase activity from HRP-conjugated antibodies and probes in Western blotting.
SuperSignal West Femto Substrate Maximum Sensitivity Substrate is an ultra-sensitive chemiluminescent substrate system for Western blotting with horseradish peroxidase (HRP) enzyme. The substrate kit enables detection of low femtogram (that's mid-zeptomole!) amounts of protein deposited on nitrocellulose or PVDF membrane and probed with appropriate primary and secondary antibodies. When combined with optimized antibody concentrations and blocking buffers, SuperSignal West Femto Substrate enables detection of target proteins in amounts that are small to be seen with typical ECL substrates.Highlights:
ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
Sensitive – detect low-femtogram (mid-zeptomole) amounts of protein in bands on nitrocellulose or PVDF membranes
Quantitative – produces quantitative signal that is measurable over two orders of magnitude
Saves on antibody – optimized for very dilute antibody concentrations; dilute primary antibodies 5000- to 100,000-fold and secondary antibodies 100,000 to 500,000-fold (from 1mg/mL stocks)
Intense signal – easy to capture an image by exposure to autoradiography film or via a CCD camera or similar imaging system
Excellent signal duration – 8 hours of usable light-output when conditions are optimized
Stable reagent – kit components are stable for 1 year at 4°C or 6 months at room temperature
Specifications
Substrate | |
Liquid | |
Horseradish Peroxidase |
Chemiluminescent | |
SuperSignal™ | |
6 to 8 hr. |
For Research Use Only. Not for use in diagnostic procedures.