Sonoco™ ThermoSafe Insulated Storage and Transport Chests
Dry ice storage chests provide mobile, non-electrical low-temperature maintenance for a wide variety of laboratory and field applications. Ideal for dry ice or wet ice storage, hot storage, or field collection.
Supplier:  Sonoco™ 390
 View more versions of this product
Catalog No. 03-530-27
Description
Description
Ideal for dry ice and wet ice storage, short-term frozen and hot storage.
  • Efficient, non deteriorating urethane foam insulation
  • Resist vapor, water, rust, corrosion
  • Washable
  • Interior corners are rounded for easy cleanup
  • Lid is hinged (except 50 lb. size) and includes support that keeps it upright
Order Info
Dolly not included with chests
Specifications
Specifications
35.07 cm
54.12 cm
37.79 cm
21.31 x 14.88 x 13.81 in. (54.12 x 37.79 x 35.07 cm)
1.2 cu. ft.
Lid Hindge w/ stay, 2 latches, and carrying handle
16 lb.
10 days
Polyethylene
1.0 cu. ft.
13.81 in.
21.31 in.
14.88 in.
Hinged
Sonoco™ ThermoSafe Aluminum Dollies (Fisher Cat. 11-676-24)
Ideal for dry ice and wet ice storage, short-term frozen and hot storage
2.25 in.
Carrying Handle
Hard-Cased Transporter