Honeywell™ Uvex Avatar™ OTG Safety Glasses
OTG (Over-the-Glass) safety glasses have a stylish ultra-light design with Weight Distribution Technology.
Supplier: Honeywell™ S3852
Description
Polycarbonate Lens Coatings available in:
- Anti-Reflective coating
- Hard coat
Fit
- Built with MMT – Multi-Material Technology™ Plus providing a high level of adjustability
- Unique Weight Distribution Technology
- 9-base wrap-around lens provides uncompromised peripheral vision and protection
- Ratchet lens: angle-adjustable temple length and lens inclination of 15 degrees
- Soft, wire-core temples to customize to any facial profile
- Flexible and 3-point adjustable nose pads to fit any nasal profile and reduce slippage
- Soft brow frame with indirect venting
- Frames available in three frame colors
Protection
- Meets ANSI Z87.1 standards and CSA Z94.3-2015 requirements.
- “U6” grade-level filtering >99.9% of UVB/UVA protection for far and near UV transmittance
- Available in 3 frame colors and 2 lens tints
Style
- ¾ frame, high performance, contemporary, stylish
Specifications
Hardcoat | |
Gray | |
Wire-core bayonet | |
Polycarbonate | |
U6 Grade |
Meets ANSI Z87.1 standards and CSA Z94.3-2015 requirements. | |
Red | |
Black | |
PC/TPU |