CAS RN 1122-28-7
CAS RN 1122-28-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
2
4,5-Dicyanoimidazole, 99%
CAS: 1122-28-7 Molecular Formula: C5H2N4 Molecular Weight (g/mol): 118.10 MDL Number: MFCD00005194 InChI Key: XGDRLCRGKUCBQL-UHFFFAOYSA-N PubChem CID: 70729 IUPAC Name: 1H-imidazole-4,5-dicarbonitrile SMILES: N#CC1=C(N=CN1)C#N
3
4,5-Dicyanoimidazole, 98+%
CAS: 1122-28-7 Molecular Formula: C5H2N4 Molecular Weight (g/mol): 118.10 MDL Number: MFCD00005194 InChI Key: XGDRLCRGKUCBQL-UHFFFAOYSA-N PubChem CID: 70729 IUPAC Name: 1H-imidazole-4,5-dicarbonitrile SMILES: N#CC1=C(N=CN1)C#N
4
4,5-Dicyanoimidazole 98.0+%, TCI America™
CAS: 1122-28-7 Molecular Formula: C5H2N4 Molecular Weight (g/mol): 118.10 MDL Number: MFCD00005194 InChI Key: XGDRLCRGKUCBQL-UHFFFAOYSA-N PubChem CID: 70729 IUPAC Name: 1H-imidazole-4,5-dicarbonitrile SMILES: N#CC1=C(N=CN1)C#N