accessibility menu, dialog, popup

UserName

CAS RN 132-66-1

OHOONH

CAS RN 132-66-1

IUPAC Name: 2-[(naphthalen-1-yl)carbamoyl]benzoic acid
Synonyms: naptalam N-1-napthylphthalamic acid N-1-naphthyl-phthalamidsaeure NPA α-naphthylphthalamic acid npa (naptalam)
Molecular Weight (g/mol): 291.31
Molecular Formula: C18H13NO3
InChi Key: JXTHEWSKYLZVJC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C(=O)NC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results