accessibility menu, dialog, popup

UserName

CAS RN 14648-57-8

H3COH

CAS RN 14648-57-8

IUPAC Name: 2-ethyladamantan-2-ol
Synonyms: 2-ethyladamantan-2-ol
Molecular Weight (g/mol): 180.29
Molecular Formula: C12H20O
InChi Key: YUBKBLFRGDNIDR-UHFFFAOYSA-N
SMILES: CCC1(O)C2CC3CC(C2)CC1C3
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results