CAS RN 1551-44-6
CAS RN 1551-44-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyclohexyl Butyrate 98.0+%, TCI America™
CAS: 1551-44-6 Molecular Formula: C10H18O2 Molecular Weight (g/mol): 170.252 MDL Number: MFCD00046354 InChI Key: VZHUBBUZNIULNM-UHFFFAOYSA-N Synonym: cyclohexyl butyrate,butanoic acid, cyclohexyl ester,cyclohexanyl butyrate,butyric acid, cyclohexyl ester,cyclohexyl n-butyrate,unii-fhh53z3i16,fema no. 2351,ccris 6553,cyclohexanol butanoate,cyclohexyl-n-butyrate PubChem CID: 243783 IUPAC Name: cyclohexyl butanoate SMILES: CCCC(=O)OC1CCCCC1