CAS RN 161599-46-8
CAS RN 161599-46-8
IUPAC Name:
4-(acetyloxy)-5-(4-amino-5-fluoro-2-oxo-1,2-dihydropyrimidin-1-yl)-2-methyloxolan-3-yl acetate
Synonyms:
4-(acetyloxy)-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl acetate
Molecular Weight (g/mol):
329.28
Molecular Formula:
C13H16FN3O6
InChi Key:
NWJBWNIUGNXJGO-UHFFFAOYNA-N
SMILES:
CC1OC(C(OC(C)=O)C1OC(C)=O)N1C=C(F)C(N)=NC1=O
1
–
1
of
1
results
1
2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine 98.0+%, TCI America™
CAS: 161599-46-8 Molecular Formula: C13H16FN3O6 Molecular Weight (g/mol): 329.284 MDL Number: MFCD07369302 InChI Key: NWJBWNIUGNXJGO-RPULLILYSA-N PubChem CID: 11809635 IUPAC Name: [(2R,3R,4R,5R)-4-acetyloxy-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl] acetate SMILES: CC1C(C(C(O1)N2C=C(C(=NC2=O)N)F)OC(=O)C)OC(=O)C