accessibility menu, dialog, popup

UserName

CAS RN 16867-04-2

OHHNO

CAS RN 16867-04-2

IUPAC Name: 3-hydroxy-1,2-dihydropyridin-2-one
Synonyms: 3-hydroxy-1H-pyridin-2-one
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: GGOZGYRTNQBSSA-UHFFFAOYSA-N
SMILES: OC1=CC=CNC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results