CAS RN 19056-41-8
CAS RN 19056-41-8
IUPAC Name:
3-bromo-4-methoxyaniline
Synonyms:
3-bromo-4-methoxyaniline
Molecular Weight (g/mol):
202.05
Molecular Formula:
C7H8BrNO
InChi Key:
NMUFTXMBONJQTC-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C=C(N)C=C1
1
–
1
of
1
results
1
3-Bromo-4-methoxyaniline 96.0+%, TCI America™
CAS: 19056-41-8 Molecular Formula: C7H8BrNO Molecular Weight (g/mol): 202.051 MDL Number: MFCD04094339 InChI Key: NMUFTXMBONJQTC-UHFFFAOYSA-N Synonym: 4-Amino-2-bromoanisole PubChem CID: 961083 IUPAC Name: 3-bromo-4-methoxyaniline SMILES: COC1=C(C=C(C=C1)N)Br