CAS RN 21190-88-5
CAS RN 21190-88-5
IUPAC Name:
ethyl 6-bromopyridine-2-carboxylate
Synonyms:
ethyl 6-bromopyridine-2-carboxylate
Molecular Weight (g/mol):
230.06
Molecular Formula:
C8H8BrNO2
InChi Key:
OONBOXSIWCBAHZ-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=NC(Br)=CC=C1
1
–
1
of
1
results
1
Ethyl 6-Bromopyridine-2-carboxylate 98.0+%, TCI America™
CAS: 21190-88-5 Molecular Formula: C8H8BrNO2 Molecular Weight (g/mol): 230.061 MDL Number: MFCD04116913 InChI Key: OONBOXSIWCBAHZ-UHFFFAOYSA-N Synonym: 6-Bromopicolinic Acid Ethyl Ester, Ethyl 6-Bromopicolinate, 6-Bromopyridine-2-carboxylic Acid Ethyl Ester PubChem CID: 2758810 IUPAC Name: ethyl 6-bromopyridine-2-carboxylate SMILES: CCOC(=O)C1=NC(=CC=C1)Br