accessibility menu, dialog, popup

UserName

CAS RN 2124-47-2

CH3CH3H2NNOO

CAS RN 2124-47-2

IUPAC Name: 2,4-dimethyl-5-nitroaniline
Synonyms: 2,4-dimethyl-5-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: DUIOVGPUAJSYCD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1N)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results