CAS RN 213178-97-3
CAS RN 213178-97-3
IUPAC Name:
2-acetamido-3-(1H-imidazol-5-yl)propanoic acid hydrate
Synonyms:
2-acetamido-3-(3H-imidazol-4-yl)propanoic acid hydrate
Molecular Weight (g/mol):
215.21
Molecular Formula:
C8H13N3O4
InChi Key:
PSWSDQRXCOJSFC-UHFFFAOYNA-N
SMILES:
O.CC(=O)NC(CC1=CN=CN1)C(O)=O
1
–
1
of
1
results
1
N-Acetyl-DL-histidine Monohydrate 98.0+%, TCI America™
CAS: 213178-97-3 Molecular Formula: C8H13N3O4 Molecular Weight (g/mol): 215.209 MDL Number: MFCD00065961 InChI Key: PSWSDQRXCOJSFC-UHFFFAOYSA-N Synonym: 2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate,ac-dl-his-oh.h2o,n-acetyl-dl-histidine monohydrate,2-acetamido-3-1h-imidazol-5-yl propanoic acid hydrate,2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate,n-acetyl-l-histidine monohydrate,2-acetamido-3-3h-imidazol-4-yl propanoic acid hydrate,acmc-1c6iz,acmc-209j3m,n-acetyl-dl-histidine hydrate PubChem CID: 18668507 IUPAC Name: 2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate SMILES: CC(=O)NC(CC1=CN=CN1)C(=O)O.O