CAS RN 28141-13-1
CAS RN 28141-13-1
IUPAC Name:
1-ethyl-2-hydroxy-4-methyl-6-oxo-1,6-dihydropyridine-3-carbonitrile
Synonyms:
1-ethyl-2-hydroxy-4-methyl-6-oxopyridine-3-carbonitrile
Molecular Weight (g/mol):
178.19
Molecular Formula:
C9H10N2O2
InChi Key:
YSNMMQRIPFUHAO-UHFFFAOYSA-N
SMILES:
CCN1C(=O)C=C(C)C(C#N)=C1O
1
–
1
of
1
results
1
3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone 98.0+%, TCI America™
CAS: 28141-13-1 Molecular Formula: C9H10N2O2 Molecular Weight (g/mol): 178.19 MDL Number: MFCD00142624 InChI Key: YSNMMQRIPFUHAO-UHFFFAOYSA-N Synonym: 3-Cyano-1-ethyl-6-hydroxy-3-picoline, 1-Ethyl-1,2-dihydro-6-hydroxy-4-methyl-2-oxo-3-pyridinecarbonitrile PubChem CID: 119879 IUPAC Name: 1-ethyl-2-hydroxy-4-methyl-6-oxo-1,6-dihydropyridine-3-carbonitrile SMILES: CCN1C(=O)C=C(C)C(C#N)=C1O