accessibility menu, dialog, popup

UserName

CAS RN 284462-37-9

CH3NHONOH2N

CAS RN 284462-37-9

IUPAC Name: 4-(4-aminophenoxy)-N-methylpyridine-2-carboxamide
Synonyms: 4-(4-aminophenoxy)-N-methylpyridine-2-carboxamide
Molecular Weight (g/mol): 243.27
Molecular Formula: C13H13N3O2
InChi Key: RXZZBPYPZLAEFC-UHFFFAOYSA-N
SMILES: CNC(=O)C1=NC=CC(OC2=CC=C(N)C=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results