accessibility menu, dialog, popup

UserName

CAS RN 287952-67-4

NHOOFFF

CAS RN 287952-67-4

IUPAC Name: 4-[4-(trifluoromethoxy)phenoxy]piperidine
Synonyms: 4-[4-(trifluoromethoxy)phenoxy]piperidine
Molecular Weight (g/mol): 261.24
Molecular Formula: C12H14F3NO2
InChi Key: RPQOTFPZKNHYFB-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(OC2CCNCC2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results