accessibility menu, dialog, popup

UserName

CAS RN 288067-35-6

BrHON

CAS RN 288067-35-6

IUPAC Name: 4-bromo-2-hydroxybenzonitrile
Synonyms: 4-bromo-2-hydroxybenzonitrile
Molecular Weight (g/mol): 198.02
Molecular Formula: C7H4BrNO
InChi Key: PAHSHGVACWNGEY-UHFFFAOYSA-N
SMILES: OC1=C(C=CC(Br)=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results