accessibility menu, dialog, popup

UserName

CAS RN 30736-07-3

H3CCH3H3CSICH3CH3H3C

CAS RN 30736-07-3

IUPAC Name: tert-butyl(tert-butylsulfanyl)iodanuide
Synonyms: tert-butyl(tert-butylsulfanyl)iodanuide
Molecular Weight (g/mol): 273.2
Molecular Formula: C8H18IS
InChi Key: MDFYNXFLLPGUBM-UHFFFAOYSA-N
SMILES: CC(C)(C)S[I-]C(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results