CAS RN 338453-21-7
CAS RN 338453-21-7
IUPAC Name:
oxane-4-sulfonyl chloride
Synonyms:
oxane-4-sulfonyl chloride
Molecular Weight (g/mol):
184.63
Molecular Formula:
C5H9ClO3S
InChi Key:
QQSBPTQNEOJFBO-UHFFFAOYSA-N
SMILES:
ClS(=O)(=O)C1CCOCC1
1
–
2
of
2
results
2
Tetrahydropyran-4-sulfonyl chloride, 97%
CAS: 338453-21-7 Molecular Formula: C5H9ClO3S Molecular Weight (g/mol): 184.64 InChI Key: QQSBPTQNEOJFBO-UHFFFAOYSA-N Synonym: tetrahydro-2h-pyran-4-sulfonyl chloride,tetrahydropyran-4-sulfonyl chloride,tetrahydro-pyran-4-sulfonyl chloride,2h-pyran-4-sulfonylchloride, tetrahydro,tetrahydropyran-4-sulfonylchloride,tetrahydropyran-4-sulphonyl chloride,2h-pyran-4-sulfonylchloride,tetrahydro,n-5-bromo-2-methoxybenzyl cyclohexanamine hydrobromide PubChem CID: 22637030 IUPAC Name: oxane-4-sulfonyl chloride SMILES: C1COCCC1S(=O)(=O)Cl