CAS RN 3970-13-6
CAS RN 3970-13-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benzyl Chloromethyl Sulfide 96.0+%, TCI America™
CAS: 3970-13-6 Molecular Formula: C8H9ClS Molecular Weight (g/mol): 172.67 MDL Number: MFCD02093436 InChI Key: UMFDSFPDTLREFS-UHFFFAOYSA-N Synonym: alpha-(Chloromethylthio)toluene PubChem CID: 319930 IUPAC Name: chloromethylsulfanylmethylbenzene SMILES: C1=CC=C(C=C1)CSCCl