CAS RN 39795-60-3
CAS RN 39795-60-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(4-Bromophenyl)pyridine 98.0+%, TCI America™
CAS: 39795-60-3 Molecular Formula: C11H8BrN Molecular Weight (g/mol): 234.096 MDL Number: MFCD00956748 InChI Key: GYJBDJGUNDKZKO-UHFFFAOYSA-N PubChem CID: 616283 IUPAC Name: 4-(4-bromophenyl)pyridine SMILES: C1=CC(=CC=C1C2=CC=NC=C2)Br