accessibility menu, dialog, popup

UserName

CAS RN 41404-58-4

FBrN

CAS RN 41404-58-4

IUPAC Name: 2-bromo-5-fluoropyridine
Synonyms: 2-bromo-5-fluoropyridine
Molecular Weight (g/mol): 175.99
Molecular Formula: C5H3BrFN
InChi Key: UODINHBLNPPDPD-UHFFFAOYSA-N
SMILES: FC1=CC=C(Br)N=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)