accessibility menu, dialog, popup

UserName

CAS RN 452972-08-6

BNBrHOOH

CAS RN 452972-08-6

IUPAC Name: (2-bromopyridin-3-yl)boronic acid
Synonyms: 2-bromopyridin-3-ylboronic acid
Molecular Weight (g/mol): 201.81
Molecular Formula: C5H5BBrNO2
InChi Key: ODWRFKJNUJLAHM-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Br)N=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results