CAS RN 4587-33-1
CAS RN 4587-33-1
IUPAC Name:
4-nitrophenyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-carbamoylpropanoate
Synonyms:
4-nitrophenyl (2R)-2-[(tert-butoxycarbonyl)amino]-3-carbamoylpropanoate
Molecular Weight (g/mol):
353.33
Molecular Formula:
C15H19N3O7
InChi Key:
IAPXDJMULQXGDD-LLVKDONJSA-N
SMILES:
CC(C)(C)OC(=O)N[C@H](CC(N)=O)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Nalpha-(tert-Butoxycarbonyl)-L-asparagine 4-Nitrophenyl Ester 98.0+%, TCI America™
CAS: 4587-33-1 Molecular Formula: C15H19N3O7 Molecular Weight (g/mol): 353.331 MDL Number: MFCD00056109 InChI Key: IAPXDJMULQXGDD-NSHDSACASA-N Synonym: boc-asn-onp,boc-l-asparagine 4-nitrophenyl ester,boc-asn-ophno2,s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate,4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate,pubchem12095,threonin-tert.-butylether,ch3 3coconhch cooh ch2cooh,n1-t-boc c1-p-nitrophenol asparagine,boc-asn-onp hplc PubChem CID: 439663 ChEBI: CHEBI:3147 IUPAC Name: (4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate SMILES: CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]