CAS RN 5060-82-2
CAS RN 5060-82-2
IUPAC Name:
7-methoxynaphthalen-2-ol
Synonyms:
7-methoxynaphthalen-2-ol
Molecular Weight (g/mol):
174.2
Molecular Formula:
C11H10O2
InChi Key:
UNFNRIIETORURP-UHFFFAOYSA-N
SMILES:
COC1=CC=C2C=CC(O)=CC2=C1
1
–
2
of
2
results
1
7-Methoxy-2-naphthol, 97%
CAS: 5060-82-2 Molecular Formula: C11H10O2 Molecular Weight (g/mol): 174.20 MDL Number: MFCD00075494 InChI Key: UNFNRIIETORURP-UHFFFAOYSA-N Synonym: 7-methoxy-2-naphthol,2-naphthalenol, 7-methoxy,7-methoxy-2-naphthalenol,pubchem16354,7-methoxy-beta-naphthol,acmc-1bnbv,7-methoxy-naphthalen-2-ol,7-methyloxy-2-naphthalenol,2-hydroxy-7-methoxynaphthalene PubChem CID: 853173 IUPAC Name: 7-methoxynaphthalen-2-ol SMILES: COC1=CC=C2C=CC(O)=CC2=C1