CAS RN 50679-08-8
CAS RN 50679-08-8
IUPAC Name:
1-(4-tert-butylphenyl)-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butan-1-ol
Synonyms:
terfenadine terfenadin-ratiopharm terfenadine product terfedura triludan teldanex terfenadine (product) terfenadin von ct terfenadin AL terfenadina
Molecular Weight (g/mol):
471.69
Molecular Formula:
C32H41NO2
InChi Key:
GUGOEEXESWIERI-UHFFFAOYNA-N
SMILES:
CC(C)(C)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Terfenadine
CAS: 50679-08-8 Molecular Formula: C32H41NO2