CAS RN 51605-97-1
CAS RN 51605-97-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Bromo-4-isopropylaniline 98.0+%, TCI America™
CAS: 51605-97-1 Molecular Formula: C9H12BrN Molecular Weight (g/mol): 214.11 MDL Number: MFCD00051585 InChI Key: WEMDUNBELVTSRP-UHFFFAOYSA-N Synonym: 2-bromo-4-isopropylaniline,2-bromo-4-isopropyl-phenylamine,4-amino-2-bromocumene,benzenamine, 2-bromo-4-1-methylethyl,zlchem 981,pubchem17446,acmc-1anzo,2-bromo-4-isopropyl aniline,2-bromo4-isopropyl-phenylamine,2-bromo-4-2-propyl aniline PubChem CID: 521337 IUPAC Name: 2-bromo-4-(propan-2-yl)aniline SMILES: CC(C)C1=CC=C(N)C(Br)=C1