CAS RN 53266-94-7
CAS RN 53266-94-7
IUPAC Name:
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
Synonyms:
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
Molecular Weight (g/mol):
186.23
Molecular Formula:
C7H10N2O2S
InChi Key:
SHQNGLYXRFCPGZ-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC1=CSC(N)=N1
1
–
1
of
1
results
1
Ethyl (2-Amino-4-thiazolyl)acetate 98.0+%, TCI America™
CAS: 53266-94-7 Molecular Formula: C7H10N2O2S Molecular Weight (g/mol): 186.23 MDL Number: MFCD00005330 InChI Key: SHQNGLYXRFCPGZ-UHFFFAOYSA-N Synonym: ethyl 2-amino-4-thiazoleacetate,ethyl 2-2-aminothiazol-4-yl acetate,ethyl 2-2-amino-1,3-thiazol-4-yl acetate,ethyl 2-aminothiazole-4-acetate,ethyl 2-amino-1,3-thiazol-4-yl acetate,ethyl 2-aminothiazol-4-acetate,2-amino-4-thiazolyl acetic acid ethyl ester,4-thiazoleacetic acid, 2-amino-, ethyl ester,ethyl 2-amino-4-thiazolyl acetate PubChem CID: 104454 IUPAC Name: ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate SMILES: CCOC(=O)CC1=CSC(N)=N1