accessibility menu, dialog, popup

UserName

CAS RN 53312-79-1

NH2ClN

CAS RN 53312-79-1

IUPAC Name: 3-amino-4-chlorobenzonitrile
Synonyms: 3-amino-4-chlorobenzonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: KRBRYJLBILJHAS-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Cl)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results