CAS RN 55650-59-4
CAS RN 55650-59-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-N,N-Dimethyl-1-[(R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine 95.0+%, TCI America™
CAS: 55650-59-4 Molecular Formula: C38H37FeNP2 MDL Number: MFCD00075283 Synonym: (S)-(R)-BPPFA