CAS RN 56344-32-2
CAS RN 56344-32-2
IUPAC Name:
3-[(2-aminoethyl)amino]propan-1-ol
Synonyms:
3-[(2-aminoethyl)amino]propan-1-ol
Molecular Weight (g/mol):
118.18
Molecular Formula:
C5H14N2O
InChi Key:
KTLIZDDPOZZHCD-UHFFFAOYSA-N
SMILES:
NCCNCCCO
1
–
1
of
1
results
1
N-(3-Hydroxypropyl)ethylenediamine 97.0+%, TCI America™
CAS: 56344-32-2 Molecular Formula: C5H14N2O Molecular Weight (g/mol): 118.18 MDL Number: MFCD00059848 InChI Key: KTLIZDDPOZZHCD-UHFFFAOYSA-N Synonym: N-(2-Aminoethyl)propanolamine, 2-(3-Hydroxypropylamino)ethylamine PubChem CID: 10973740 IUPAC Name: 3-(2-aminoethylamino)propan-1-ol SMILES: C(CNCCN)CO