accessibility menu, dialog, popup

UserName

CAS RN 5678-45-5

H3CO(R)H3NOO

CAS RN 5678-45-5

IUPAC Name: (3R)-3-azaniumyl-3-(4-methoxyphenyl)propanoate
Synonyms: (3R)-3-ammonio-3-(4-methoxyphenyl)propanoate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: NYTANCDDCQVQHG-SECBINFHSA-N
SMILES: COC1=CC=C(C=C1)[C@H]([NH3+])CC([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)