accessibility menu, dialog, popup

UserName

CAS RN 584-79-2

CH3CH3OOOH3CH2CH3CCH3

CAS RN 584-79-2

IUPAC Name: 2-methyl-4-oxo-3-(prop-2-en-1-yl)cyclopent-2-en-1-yl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate
Synonyms: pyrethrin caswell no. 025 necarboxylic acid allyl cinerins allethrine allethrins allyl cinerin I bioaletrina pynamin esbiol TM intermediate 3336
Molecular Weight (g/mol): 302.41
Molecular Formula: C19H26O3
InChi Key: ZCVAOQKBXKSDMS-UHFFFAOYNA-N
SMILES: CC(C)=CC1C(C(=O)OC2CC(=O)C(CC=C)=C2C)C1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results