CAS RN 61204-00-0
CAS RN 61204-00-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(trans-4-Butylcyclohexyl)benzonitrile 98.0+%, TCI America™
CAS: 61204-00-0 Molecular Formula: C17H23N Molecular Weight (g/mol): 241.38 MDL Number: MFCD00799425 InChI Key: YYAVXASAKUOZJJ-UHFFFAOYSA-N Synonym: 4-trans-4-butylcyclohexyl benzonitrile,4-4-butylcyclohexyl benzonitrile,p-4-butylcyclohexyl benzonitrile,benzonitrile, 4-4-butylcyclohexyl,benzonitrile, 4-trans-4-butylcyclohexyl,4-trans-4-butylcyclohexyl-benzonitrile,trans-4-4'-n-butylcyclohexyl-benzonitrile,benzonitrile,4-trans-4-butylcyclohexyl,4-4-butylcyclohexyl benzenecarbonitrile,4-1s,4r-4-butylcyclohexyl benzonitrile PubChem CID: 109062 IUPAC Name: 4-(4-butylcyclohexyl)benzonitrile SMILES: CCCCC1CCC(CC1)C1=CC=C(C=C1)C#N