accessibility menu, dialog, popup

UserName

CAS RN 6147-11-1

CH3OHOOOHCH3CH3OHOCH3H3C

CAS RN 6147-11-1

IUPAC Name: 1,3,6-trihydroxy-7-methoxy-2,8-bis(3-methylbut-2-en-1-yl)-9H-xanthen-9-one
Synonyms: mangostin α-mangostin
Molecular Weight (g/mol): 410.47
Molecular Formula: C24H26O6
InChi Key: GNRIZKKCNOBBMO-UHFFFAOYSA-N
SMILES: COC1=C(O)C=C2OC3=CC(O)=C(CC=C(C)C)C(O)=C3C(=O)C2=C1CC=C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results