CAS RN 620-81-5
CAS RN 620-81-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Oxanilide 98.0+%, TCI America™
CAS: 620-81-5 Molecular Formula: C14H12N2O2 Molecular Weight (g/mol): 240.26 MDL Number: MFCD00041758 InChI Key: FTWUXYZHDFCGSV-UHFFFAOYSA-N Synonym: oxanilide,oxanilid,oxalanilide,n,n'-diphenylethanediamide,ethanediamide, n,n'-diphenyl,n,n'-diphenyloxalamide,oxaldianilide,oxamide, n,n'-diphenyl,ethanediamide, n1,n2-diphenyl,oxanilide 6ci,7ci,8ci PubChem CID: 69288 IUPAC Name: N,N'-diphenylethanediamide SMILES: O=C(NC1=CC=CC=C1)C(=O)NC1=CC=CC=C1