accessibility menu, dialog, popup

UserName

CAS RN 62621-09-4

CH3NOOOOCH3

CAS RN 62621-09-4

IUPAC Name: methyl 2-methyl-4-nitrobenzoate
Synonyms: methyl 2-methyl-4-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: JJHCLPDHYBSSHC-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results